Ammoniumhydrogencarbonat ≥99,0%, ReagentPlus®
Lieferant: Merck
|
Achtung
|
Synonyme:
Ammoniumbicarbonat
A6141-25G
SIALA6141-25GEA
38
EUR
SIALA6141-25G
Ammoniumhydrogencarbonat ≥99,0%, ReagentPlus®
Ammoniumhydrogencarbonat
Formel:
NH₄HCO₃ Molecular Weight: 79,06 g/mol Schmelzpunkt: 35 °C Dichte: 1,57 g/cm³ (20 °C) |
MDL:
MFCD00012138 CAS-Nummer: 1066-33-7 EINECS: 213-911-5 Merck Index: 13,00497 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...