Agmatine sulphate ≥97%, Pulver, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulphate

M-Clarity™ Qualitätsstufe = MQ100

A7127-1G
SIALA7127-1GEA 158 EUR
SIALA7127-1G
Agmatine sulphate ≥97%, Pulver, Sigma-Aldrich®
Agmatine sulphate

Putative endogenous neurotransmitter at imidazoline receptors; displaces clonidine at α2-adrenergic and at imidazoline receptors; blocks NMDA-activated ion channels in hippocampal neurons.

Formel: C₅H₁₄N₄
Molecular Weight: 228,27 g/mol
Schmelzpunkt: 234…238 °C (Lit.)
MDL: MFCD00013109
CAS-Nummer: 2482-00-0

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Off-White
Appearance (Form) Powder
Infrared spectrum ≥97%
Solubility (Turbidity)(50 mg/ml in H₂O) Clear to Slightly Hazy
Solubility (Color) Colorless to Faint Yellow

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR