N-(2-Acetamido)-2-aminoethansulfonsäure (ACES), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
A7949-100G
A7949-250G
A7949-25G
SIALA7949-100GEA
605
EUR
SIALA7949-100G
SIALA7949-250G
SIALA7949-25G
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES), Sigma-Aldrich®
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES)
Formel:
H₂NCOCH₂NHCH₂CH₂SO₃H Schmelzpunkt: 270…295 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008030 CAS-Nummer: 7365-82-4 EINECS: 230-908-4 Merck Index: 13,00037 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...