N-Aminorhodanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-2-thioxo-1,3-thiazolidin-4-one, 3-Aminorhodanine

A79507-10G
SIALA79507-10GEA 72.1 EUR
SIALA79507-10G
N-Aminorhodanin, Sigma-Aldrich®
N-Aminorhodanin
Formel: C₃H₄N₂OS₂
Molecular Weight: 148,21 g/mol
MDL: MFCD00005489
CAS-Nummer: 1438-16-0
EINECS: 215-870-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR