α-Angelicalacton, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Methylfuran-2(3H)-on
M-Clarity™ Qualitätsstufe = MQ100
A86406-100G
A86406-10G
SIALA86406-100GEA
190
EUR
SIALA86406-100G
SIALA86406-10G
α-Angelicalacton, Sigma-Aldrich®
α-Angelicalacton
Formel:
C₅H₆O₂ Molecular Weight: 98.1 g/mol Siedepunkt: 249 Schmelzpunkt: 0 Dichte: 1.0904 g/cm³ (12 °C) |
MDL:
MFCD00005375 CAS-Nummer: 591-12-8 EINECS: 209-701-8 Merck Index: 12,00686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...