Adenin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Purin-6-amin Hydrochlorid, 1H-Adenin Hydrochlorid, 9H-Purin-6-amin Hydrochlorid, 6-Aminopurin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ200

A8751-1G A8751-100G A8751-25G A8751-5G
SIALA8751-1GEA 29.6 EUR
SIALA8751-1G SIALA8751-100G SIALA8751-25G SIALA8751-5G
Adenin Hydrochlorid, Sigma-Aldrich®
Adenin Hydrochlorid
Formel: C₅H₅N₅·HCl
Molecular Weight: 171.59 g/mol
MDL: MFCD00150864
CAS-Nummer: 2922-28-3
EINECS: 220-867-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR