2,6-Dihydroxy-9,10-anthrachinon, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2,6-Dihydroxyanthrachinon
M-Clarity™ Qualitätsstufe = MQ100
A89502-5G
SIALA89502-5GEA
178
EUR
SIALA89502-5G
2,6-Dihydroxy-9,10-anthrachinon, Sigma-Aldrich®
2,6-Dihydroxy-9,10-anthrachinon
Formel:
C₁₄H₈O₄ Molecular Weight: 240,22 g/mol Schmelzpunkt: > 330 °C |
MDL:
MFCD00001228 CAS-Nummer: 84-60-6 EINECS: 201-544-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...