Adeninsulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Adeninhemisulfat

A9126-100G A9126-25G A9126-1KG
SIALA9126-100GEA 546 EUR
SIALA9126-100G SIALA9126-25G SIALA9126-1KG
Adeninsulfat, Sigma-Aldrich®
Adeninsulfat
Formel: C₅H₅N₅·½H₂SO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00213655
CAS-Nummer: 321-30-2
EINECS: 206-286-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR