Ampicillin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

A9393-5G A9393-25G
SIALA9393-5GEA 75.7 EUR
SIALA9393-5G SIALA9393-25G
Ampicillin, Sigma-Aldrich®
Ampicillin
Formel: C₁₆H₁₉N₃O₄S
Molecular Weight: 349.4 g/mol
MDL: MFCD00005175
CAS-Nummer: 69-53-4
EINECS: 200-709-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR