2-Aminobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung

A9628-100MG A9628-500MG A9628-1G
SIALA9628-100MGEA 94.5 EUR
SIALA9628-100MG SIALA9628-500MG SIALA9628-1G
2-Aminobenzaldehyd, Sigma-Aldrich®
2-Aminobenzaldehyd
Formel: C₇H₇NO
Molecular Weight: 121.14 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00007709
CAS-Nummer: 529-23-7
EINECS: 208-454-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR