2-Aminobenzaldehyd, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
A9628-100MG
A9628-500MG
A9628-1G
SIALA9628-100MGEA
94.5
EUR
SIALA9628-100MG
SIALA9628-500MG
SIALA9628-1G
2-Aminobenzaldehyd, Sigma-Aldrich®
2-Aminobenzaldehyd
Formel:
C₇H₇NO Molecular Weight: 121.14 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00007709 CAS-Nummer: 529-23-7 EINECS: 208-454-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...