Bortrifluorid-Methanol-Komplex 14% in Methanol, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
B1252-100ML
B1252-1L
B1252-250ML
B1252-500ML
B1252-5ML
SIALB1252-100MLEA
77.8
EUR
SIALB1252-100ML
SIALB1252-1L
SIALB1252-250ML
SIALB1252-500ML
SIALB1252-5ML
Bortrifluorid-Methanol-Komplex 14% in Methanol, Sigma-Aldrich®
Bortrifluorid-Methanol-Komplex
Formel:
CH₄BF₃O |
MDL:
MFCD00071635 CAS-Nummer: 373-57-9 EINECS: 206-766-4 UN: 3286 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...