Bortrifluorid-Methanol-Komplex 14% in Methanol, Sigma-Aldrich®

Lieferant: Merck
Gefahr

B1252-100ML B1252-1L B1252-250ML B1252-500ML B1252-5ML
SIALB1252-100MLEA 77.8 EUR
SIALB1252-100ML SIALB1252-1L SIALB1252-250ML SIALB1252-500ML SIALB1252-5ML
Bortrifluorid-Methanol-Komplex 14% in Methanol, Sigma-Aldrich®
Bortrifluorid-Methanol-Komplex
Formel: CH₄BF₃O
MDL: MFCD00071635
CAS-Nummer: 373-57-9
EINECS: 206-766-4
UN: 3286
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR