2-Benzoylthiophen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Phenyl-2-thienylketon

M-Clarity™ Qualitätsstufe = MQ200

B14809-5G
SIALB14809-5GEA 28.1 EUR
SIALB14809-5G
2-Benzoylthiophen, Sigma-Aldrich®
2-Benzoylthiophen
Formel: C₁₁H₈OS
Molecular Weight: 188,25 g/mol
MDL: MFCD00022495
CAS-Nummer: 135-00-2
EINECS: 205-169-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR