BCIP/NBT, flüssiges Substrat anwendungsbereit, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
BCIP (5-bromo-4-chloro-3-indolylphosphate toluidine salt), NBT (nitro-blue tetrazolium chloride)
M-Clarity™ Qualitätsstufe = MQ200
B1911-100ML
SIALB1911-100MLEA
121
EUR
SIALB1911-100ML
BCIP/NBT, flüssiges Substrat anwendungsbereit, Sigma-Aldrich®
BCIP/NBT
BCIP/NBT liquid ready-to-use precipitating substrate system is an insoluble substrate for alkaline phosphatase.
Immunohistochemical staining with BCIP/NBT utilised requires organic mounting media and can be counterstained with nuclear fast red or light green.
Bitte beachten Not recommended for Elisa micro-well procedures.
Spezifikation Testergebnisse
Appearance (Turbidity) | Clear |
Appearance (Color) | Faint Green-Yellow to Light Green-Yellow to Light Yellow |
Appearance (Form) | Liquid |
pH | 8.5 - 9.5 |
BCIP Immunoblot Test | Compares (Compares to Powdered Substrate Mixture) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...