N-Benzylglycinethylester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-(benzylamino)acetate

M-Clarity™ Qualitätsstufe = MQ200

B22704-50G
SIALB22704-50GEA 116 EUR
SIALB22704-50G
N-Benzylglycinethylester, Sigma-Aldrich®
N-Benzylglycinethylester
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,25 g/mol
MDL: MFCD00009174
CAS-Nummer: 6436-90-4
EINECS: 229-218-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR