4′-Hydroxy-2-phenylacetophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Benzyl 4-hydroxyphenyl ketone, 1-(4-Hydroxyphenyl)-2-phenylethanone
M-Clarity™ Qualitätsstufe = MQ100
B23077-5G
SIALB23077-5GEA
78.9
EUR
SIALB23077-5G
4′-Hydroxy-2-phenylacetophenon, Sigma-Aldrich®
4′-Hydroxy-2-phenylacetophenon
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,24 g/mol |
MDL:
MFCD00002360 CAS-Nummer: 2491-32-9 EINECS: 219-654-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...