Dikaliumbenzol-1,3-disulfonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

B3159-10G
SIALB3159-10GEA 162 EUR
SIALB3159-10G
Dikaliumbenzol-1,3-disulfonat, Sigma-Aldrich®
Dikaliumbenzol-1,3-disulfonat
Formel: C₆H₄K₂O₆S₂
Molecular Weight: 314,42 g/mol
CAS-Nummer: 5710-54-3
EINECS: 227-199-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR