X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
B4252-100MG
B4252-1G
B4252-250MG
B4252-2G
B4252-500MG
B4252-50MG
B4252-5G
SIALB4252-100MGEA
104
EUR
SIALB4252-100MG
SIALB4252-1G
SIALB4252-250MG
SIALB4252-2G
SIALB4252-500MG
SIALB4252-50MG
SIALB4252-5G
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid), Sigma-Aldrich®
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Formel:
C₁₄H₁₅BrClNO₆ Schmelzpunkt: 237…239 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005666 CAS-Nummer: 7240-90-6 EINECS: 230-640-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...