BIS-TRIS-Propan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2'-(Propan-1,3-diyldiimino)bis[2-(hydroxymethyl)propan-1,3-diol], 1,3-Bis(tris(hydroxymethyl)methylamino)propan

M-Clarity™ Qualitätsstufe = MQ400

B4679-100G B4679-25G B4679-500G
SIALB4679-100GEA 385 EUR
SIALB4679-100G SIALB4679-25G SIALB4679-500G
BIS-TRIS-Propan, Sigma-Aldrich®
BIS-TRIS-Propan
Formel: C₁₁H₂₆N₂O₆
MDL: MFCD00004689
CAS-Nummer: 64431-96-5
EINECS: 264-899-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR