1,2-Di(2-pyridyl)ethylen, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
1,2-Bis(2-pyridyl)ethylene
B52603-2.5G
SIALB52603-2.5GEA
134
EUR
SIALB52603-2.5G
1,2-Di(2-pyridyl)ethylen, Sigma-Aldrich®
1,2-Di(2-pyridyl)ethylen
Formel:
C₁₂H₁₀N₂ Molecular Weight: 182,22 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 119 °C |
MDL:
MFCD00006356 CAS-Nummer: 1437-15-6 EINECS: 215-866-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...