Bradfords Reagenz für die quantitative Proteinbestimmung or 0,1 - 1,4 mg/ml protein, Supelco®
Lieferant: Merck
|
|
M-Clarity™ Qualitätsstufe = MQ200
B6916-500ML
SIALB6916-500MLEA
154
EUR
SIALB6916-500ML
Bradfords Reagenz für die quantitative Proteinbestimmung or 0,1 - 1,4 mg/ml protein, Supelco®
Bradfords Reagenz
Bradford assay is addition of coomassie brilliant blue G-250 to protein solution. The coomassie blue dye associates with basic and aromatic amino acids, thereby causing shift in absorbance during protein determination.
- The reagent is ready to use and no mixing or dilution required
- Colour development is rapid
- Only a five minute incubation and then the sample is read a 595 nm
- Reducing sugars and reducing substances along with thiols do not interfere with this reagent
- Can be used in microwell plate assays
- Reagent is suitable for micro (1 ot 10 μg/ml) and standard (50 to 1400 μg/ml) assay
Synonym: Coomassie dye binding protein assay, protein dye reagent.
UN:
1805 ADR: 8,III |
Spezifikation Testergebnisse
Appearance (Color) | Faint Yellow-Brown to Light Brown |
Appearance (Form) | Solution |
Suitability, Suitable for protein determination | Suitable |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...