1-Brom-2-naphthoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Bromonaphthalene-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

B7252-1G
SIALB7252-1GEA 39.2 EUR
SIALB7252-1G
1-Brom-2-naphthoesäure, Sigma-Aldrich®
1-Brom-2-naphthoesäure
Formel: C₁₁H₇BrO₂
Molecular Weight: 251.0800 g/mol
MDL: MFCD00021408
CAS-Nummer: 20717-79-7
EINECS: 243-984-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR