X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cyclohexylammoniumsalz der 5-Brom-4-chlor-3-indolyl-β-D-glucuronsäure, X-Glucuro CHAsalz, Cyclohexylammoniumsalz der (2S,3S,4S,5R)-6-[(5-Brom-4-chlor-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxan-2-carbonsäure
M-Clarity™ Qualitätsstufe = MQ200
B8174-5TAB
SIALB8174-5TABEA
318
EUR
SIALB8174-5TAB
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA), Sigma-Aldrich®
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)
Formel:
C₂₀H₂₆BrClN₂O₇ Molecular Weight: 521,79 g/mol Schmelzpunkt: 230 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00058543 CAS-Nummer: 114162-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...