5-Brom-DL-tryptophan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-3-(5-bromo-1H-indol-3-yl)propanoic acid

M-Clarity™ Qualitätsstufe = MQ100

B82707-1G
SIALB82707-1GEA 322 EUR
SIALB82707-1G
5-Brom-DL-tryptophan, Sigma-Aldrich®
5-Brom-DL-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
Molecular Weight: 283,12 g/mol
Schmelzpunkt: 273…276 °C
MDL: MFCD00005648
CAS-Nummer: 6548-09-0
EINECS: 229-464-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR