Benzoinmethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone

M-Clarity™ Qualitätsstufe = MQ200

B8703-100G
SIALB8703-100GEA 195 EUR
SIALB8703-100G
Benzoinmethylether, Sigma-Aldrich®
Benzoinmethylether
Formel: C₁₅H₁₄O₂
Molecular Weight: 226,27 g/mol
MDL: MFCD00008492
CAS-Nummer: 3524-62-7
EINECS: 222-538-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR