3,3',4,4'-Benzophenontetracarbonsäuredianhydrid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Benzophenone-3,3',4,4'-tetracarboxylic dianhydride
M-Clarity™ Qualitätsstufe = MQ200
B9750-100G
B9750-500G
SIALB9750-100GEA
85.3
EUR
SIALB9750-100G
SIALB9750-500G
3,3',4,4'-Benzophenontetracarbonsäuredianhydrid, Sigma-Aldrich®
3,3',4,4'-Benzophenontetracarbonsäuredianhydrid
Formel:
C₁₇H₆O₇ |
MDL:
MFCD00005923 CAS-Nummer: 2421-28-5 EINECS: 219-348-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...