3,3',4,4'-Benzophenontetracarbonsäuredianhydrid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzophenone-3,3',4,4'-tetracarboxylic dianhydride

M-Clarity™ Qualitätsstufe = MQ200

B9750-100G B9750-500G
SIALB9750-100GEA 85.3 EUR
SIALB9750-100G SIALB9750-500G
3,3',4,4'-Benzophenontetracarbonsäuredianhydrid, Sigma-Aldrich®
3,3',4,4'-Benzophenontetracarbonsäuredianhydrid
Formel: C₁₇H₆O₇
MDL: MFCD00005923
CAS-Nummer: 2421-28-5
EINECS: 219-348-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR