Benzo[k]fluoranthen, Sigma-Aldrich®

Lieferant: Merck

BCR048R-10MG
SIALBCR048R-10MGEA 168 EUR
SIALBCR048R-10MG
Benzo[k]fluoranthen, Sigma-Aldrich®
Benzo[k]fluoranthen
Formel: C₂₀H₁₂
MDL: MFCD00046287
CAS-Nummer: 207-08-9
EINECS: 205-916-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR