Benzo[e]pyren, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
BeP
BCR050-100MG
SIALBCR050-100MGEA
586
EUR
SIALBCR050-100MG
Benzo[e]pyren, Sigma-Aldrich®
Benzo[e]pyren
Formel:
C₂₀H₁₂ Molecular Weight: 252,32 g/mol Siedepunkt: 492 °C (1013 hPa) Schmelzpunkt: 178…179 °C |
CAS-Nummer:
192-97-2 EINECS: 205-892-7 UN: 3077 ADR: 9,III Merck Index: 12,01135 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...