Benzo[ghi]perylen, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
BCR052-100MG
SIALBCR052-100MGEA
586
EUR
SIALBCR052-100MG
Benzo[ghi]perylen, Sigma-Aldrich®
Benzo[ghi]perylen
Formel:
C₂₂H₁₂ Molecular Weight: 276,34 g/mol Siedepunkt: 500 °C (1013 hPa) Schmelzpunkt: 278…280 °C |
MDL:
MFCD00004135 CAS-Nummer: 191-24-2 EINECS: 205-883-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...