Quarz, BCR® CRM 14-90 μm

Lieferant: Merck

Synonyme: Silicium(IV)oxid, Sand, weißer Quarz, Siliciumdioxid

BCR069-10G
SIALBCR069-10GEA 257 EUR
SIALBCR069-10G
Quarz, BCR® CRM 14-90 μm
Quarz

Quartz (particle size) BCR® certified Reference Material, 14-90 μm particle size

Formel: SiO₂
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230
Schmelzpunkt: 1427
Dichte: 2.622…2.645 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00217788
CAS-Nummer: 14808-60-7
EINECS: 238-878-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5273273c3b2e473c6ced4e97e18a338f3d21302c&vid=9c414ebb18d830bfa8a77b318dbbb0e6a96da07b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR