Benzo[a]fluoranthen, Sigma-Aldrich®

Lieferant: Merck

BCR097-100MG
SIALBCR097-100MGEA 586 EUR
SIALBCR097-100MG
Benzo[a]fluoranthen, Sigma-Aldrich®
Benzo[a]fluoranthen
Formel: C₂₀H₁₂
CAS-Nummer: 203-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR