Dibenzo[a,i]acridin, Sigma-Aldrich®

Lieferant: Merck

BCR152-20MG
SIALBCR152-20MGEA 323 EUR
SIALBCR152-20MG
Dibenzo[a,i]acridin, Sigma-Aldrich®
Dibenzo[a,i]acridin
Formel: C₂₁H₁₃N
Molecular Weight: 279.34 g/mol
CAS-Nummer: 226-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR