Dibenzo[a,h]acridin, Sigma-Aldrich®

Lieferant: Merck

BCR153R-10MG
SIALBCR153R-10MGEA 168 EUR
SIALBCR153R-10MG
Dibenzo[a,h]acridin, Sigma-Aldrich®
Dibenzo[a,h]acridin
Formel: C₂₁H₁₃N
Molecular Weight: 279.34 g/mol
CAS-Nummer: 226-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR