Benzo[a]acridin, Sigma-Aldrich®

Lieferant: Merck

BCR157-100MG
SIALBCR157-100MGEA 586 EUR
SIALBCR157-100MG
Benzo[a]acridin, Sigma-Aldrich®
Benzo[a]acridin
Formel: C₁₇H₁₁N
Molecular Weight: 229.28 g/mol
CAS-Nummer: 225-11-6
EINECS: 205-929-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR