Benzo[c]acridin, Sigma-Aldrich®

Lieferant: Merck

BCR158-100MG
SIALBCR158-100MGEA 586 EUR
SIALBCR158-100MG
Benzo[c]acridin, Sigma-Aldrich®
Benzo[c]acridin
Formel: C₁₇H₁₁N
CAS-Nummer: 225-51-4
EINECS: 205-930-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR