Aluminiumoxid, α-Phase, Sigma-Aldrich®

Lieferant: Merck

Synonyme: α-Tonerde

BCR169-60G
SIALBCR169-60GEA 195 EUR
SIALBCR169-60G
Aluminiumoxid, α-Phase, Sigma-Aldrich®
Aluminiumoxid, α-Phase
Formel: Al₂O₃
Molecular Weight: 101,96 g/mol
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR