2,3,3'-Trichlorbiphenyl (PCB Nr. 20), Sigma-Aldrich®
Lieferant: Merck
BCR290-25MG
SIALBCR290-25MGEA
323
EUR
SIALBCR290-25MG
2,3,3'-Trichlorbiphenyl (PCB Nr. 20), Sigma-Aldrich®
2,3,3'-Trichlorbiphenyl (PCB Nr. 20)
Formel:
C₁₂H₇Cl₃ Molecular Weight: 257.55 g/mol |
CAS-Nummer:
38444-84-7 UN: 3432 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...