Magermilchpulver, Pulver

Lieferant: Merck

BCR685-50G
SIALBCR685-50GEA 240 EUR
SIALBCR685-50G
Magermilchpulver, Pulver
Magermilchpulver
Lagertemperatur: Raumtemperatur
CAS-Nummer: 999999-99-4
EINECS: 310-127-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR