Chloroform : isoamyl alcohol (24:1) (v/v), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
C0549-1PT
C0549-1QT
SIALC0549-1PTEA
122
EUR
SIALC0549-1PT
SIALC0549-1QT
Chloroform : isoamyl alcohol (24:1) (v/v), Sigma-Aldrich®
Chloroform : isoamyl alcohol
Formel:
CHCl₃,C₅H₁₂O Lagertemperatur: Raumtemperatur |
UN:
1888 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...