Chloroform : isoamyl alcohol (24:1) (v/v), Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

C0549-1PT C0549-1QT
SIALC0549-1PTEA 122 EUR
SIALC0549-1PT SIALC0549-1QT
Chloroform : isoamyl alcohol (24:1) (v/v), Sigma-Aldrich®
Chloroform : isoamyl alcohol
Formel: CHCl₃,C₅H₁₂O
Lagertemperatur: Raumtemperatur
UN: 1888
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR