1,2-Cyclohexandiondioxim, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

C102008-10G
SIALC102008-10GEA 118 EUR
SIALC102008-10G
1,2-Cyclohexandiondioxim, Sigma-Aldrich®
1,2-Cyclohexandiondioxim
Formel: C₆H₁₀N₂O₂
Molecular Weight: 142,16 g/mol
MDL: MFCD00001663
CAS-Nummer: 492-99-9
EINECS: 207-769-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR