Cyclopropyl-4-methoxyphenylketon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cyclopropyl(4-methoxyphenyl)methanone

M-Clarity™ Qualitätsstufe = MQ100

C119806-5G
SIALC119806-5GEA 52.1 EUR
SIALC119806-5G
Cyclopropyl-4-methoxyphenylketon, Sigma-Aldrich®
Cyclopropyl-4-methoxyphenylketon
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,22 g/mol
MDL: MFCD00001296
CAS-Nummer: 7152-03-6
EINECS: 230-487-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR