α-Cyclopropylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cyclopropylphenylcarbinol, Cyclopropyl-phenylmethanol

M-Clarity™ Qualitätsstufe = MQ100

C120405-1G
SIALC120405-1GEA 38.6 EUR
SIALC120405-1G
α-Cyclopropylbenzylalkohol, Sigma-Aldrich®
α-Cyclopropylbenzylalkohol
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
MDL: MFCD00001299
CAS-Nummer: 1007-03-0
EINECS: 213-749-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR