1-(3-Chlorphenyl)biguanid Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: m-CPBG hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

C144-500MG
SIALC144-500MGEA 62 EUR
SIALC144-500MG
1-(3-Chlorphenyl)biguanid Hydrochlorid, Sigma-Aldrich®
1-(3-Chlorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₁Cl₂N₅
Molecular Weight: 248,12 g/mol
MDL: MFCD00053019
CAS-Nummer: 2113-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR