Cholinhydrogentartrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate

M-Clarity™ Qualitätsstufe = MQ200

C1629-500G
SIALC1629-500GEA 124 EUR
SIALC1629-500G
Cholinhydrogentartrat, Sigma-Aldrich®
Cholinhydrogentartrat
Formel: C₉H₁₉NO₇
Molecular Weight: 253.25 g/mol
MDL: MFCD00036332
CAS-Nummer: 87-67-2
EINECS: 201-763-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR