Coelenterazine fcp, fest, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

C3105-50UG
SIALC3105-50UGEA 172 EUR
SIALC3105-50UG
Coelenterazine fcp, fest, Sigma-Aldrich®
Coelenterazine fcp

135 times higher luminescence intensity than native coelenterazine

Formel: C₂₅H₂₄FN₃O₂
MDL: MFCD18632537
CAS-Nummer: 123437-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR