3'-Desoxyadenosin (Cordycepin), Sigma-Aldrich®

Lieferant: Merck

C3394-100MG C3394-10MG C3394-25MG
SIALC3394-100MGEA 830 EUR
SIALC3394-100MG SIALC3394-10MG SIALC3394-25MG
3'-Desoxyadenosin (Cordycepin), Sigma-Aldrich®
3'-Desoxyadenosin (Cordycepin)
Formel: C₁₀H₁₃N₅O₃
MDL: MFCD00037998
CAS-Nummer: 73-03-0
EINECS: 200-791-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR