ω-Chlor-3,4-dihydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: α-Chlor-3,4-dihydroxyacetophenon, 3,4-Dihydroxyphenacylchlorid, 2-Chlor-3',4'-dihydroxyacetophenon

M-Clarity™ Qualitätsstufe = MQ100

C34405-25G
SIALC34405-25GEA 90.7 EUR
SIALC34405-25G
ω-Chlor-3,4-dihydroxyacetophenon, Sigma-Aldrich®
ω-Chlor-3,4-dihydroxyacetophenon
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD00002200
CAS-Nummer: 99-40-1
EINECS: 202-754-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR